2-Chlor-3-cyanpyrazin

Lieferant: Thermo Scientific

MAYBKM04911.250EA 0 EUR
MAYBKM04911.250 MAYBKM04911DA
2-Chlor-3-cyanpyrazin
2-Chloro-3-cyanopyrazine
Formel: C₅H₂ClN₃
Molecular Weight: 139,5400 g/mol
MDL: MFCD00219653
CAS-Nummer: 55557-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR