2-Chlor-6-(dipropylamino)pyrazin

Lieferant: COMBI-BLOCKS

HC-6398-5G HC-6398-1G
COBBHC-6398-5GEA 303 EUR
COBBHC-6398-5G COBBHC-6398-1G
2-Chlor-6-(dipropylamino)pyrazin
2-Chloro-6-(dipropylamino)pyrazine
Formel: C₁₀H₁₆ClN₃
MDL: MFCD09878390
CAS-Nummer: 957065-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR