2-Amino-6-chlorpyrazin
Lieferant: COMBI-BLOCKS
HC-6181-25G
HC-6181-100G
HC-6181-5G
COBBHC-6181-25GEA
88.6
EUR
COBBHC-6181-25G
COBBHC-6181-100G
COBBHC-6181-5G
2-Amino-6-chlorpyrazin
2-Amino-6-chloropyrazine
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol |
MDL:
MFCD00055024 CAS-Nummer: 33332-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...