2-(Butylamino)-6-chlorpyrazin

Lieferant: COMBI-BLOCKS

HC-6186-1G HC-6186-5G
COBBHC-6186-1GEA 0 EUR
COBBHC-6186-1G COBBHC-6186-5G
2-(Butylamino)-6-chlorpyrazin
2-(Butylamino)-6-chloropyrazine
Formel: C₈H₁₂ClN₃
MDL: MFCD09800983
CAS-Nummer: 951884-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR