2-Chlor-6-(isopropylamino)pyrazin

Lieferant: COMBI-BLOCKS

HC-6176-5G HC-6176-1G
COBBHC-6176-5GEA 595 EUR
COBBHC-6176-5G COBBHC-6176-1G
2-Chlor-6-(isopropylamino)pyrazin
2-Chlor-6-(isopropylamino)pyrazin
Formel: C₇H₁₀ClN₃
MDL: MFCD09800979
CAS-Nummer: 951884-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR