2-Chlor-6-(methylamino)pyrazin
Lieferant: COMBI-BLOCKS
Synonyme:
6-Chloro-N-methylpyrazin-2-amine
HC-6374-1G
HC-6374-5G
COBBHC-6374-1GEA
53.5
EUR
COBBHC-6374-1G
COBBHC-6374-5G
2-Chlor-6-(methylamino)pyrazin
2-Chloro-6-(methylamino)pyrazine
Formel:
C₅H₆ClN₃ Molecular Weight: 143,5700 g/mol |
MDL:
MFCD09878383 CAS-Nummer: 848366-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...