2-Chlor-6-(pentylamino)pyrazin

Lieferant: COMBI-BLOCKS

HC-6178-1G HC-6178-5G
COBBHC-6178-1GEA 75.9 EUR
COBBHC-6178-1G COBBHC-6178-5G
2-Chlor-6-(pentylamino)pyrazin
2-Chloro-6-(pentylamino)pyrazine
Formel: C₉H₁₄ClN₃
MDL: MFCD09800980
CAS-Nummer: 951884-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR