2-Chlor-6-((3-methoxypropyl)amino)pyrazin
Lieferant: COMBI-BLOCKS
HC-6149-1G
HC-6149-5G
COBBHC-6149-1GEA
0
EUR
COBBHC-6149-1G
COBBHC-6149-5G
2-Chlor-6-((3-methoxypropyl)amino)pyrazin
2-Chloro-6-((3-methoxypropyl)amino)pyrazine
Formel:
C₈H₁₂ClN₃O |
MDL:
MFCD09800963 CAS-Nummer: 941294-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...