2-Chlor-6-(cyclohexylamino)pyrazin

Lieferant: COMBI-BLOCKS

HC-6179-5G HC-6179-1G
COBBHC-6179-5GEA 564 EUR
COBBHC-6179-5G COBBHC-6179-1G
2-Chlor-6-(cyclohexylamino)pyrazin
2-Chloro-6-(cyclohexylamino)pyrazine
Formel: C₁₀H₁₄ClN₃
MDL: MFCD09800981
CAS-Nummer: 642459-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR