2-Cyanpyrazin

Lieferant: FLUOROCHEM

Synonyme: Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile

CFF603.5 002755-25G 002755-100G
CHMPCFF603.5EA 268 EUR
CHMPCFF603.5 FLUO002755-25G FLUO002755-100G
2-Cyanpyrazin
2-Cyanopyrazine
Formel: C₅H₃N₃
Molecular Weight: 105,10 g/mol
MDL: MFCD00049361
CAS-Nummer: 19847-12-2
EINECS: 243-369-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR