3-Amino-2-pyrazincarbonsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-2-pyrazinecarboxylic acid

B23707.06 B23707.14
B23707.06EA 32 EUR
B23707.06 B23707.14
3-Amino-2-pyrazincarbonsäure ≥98%
3-Aminopyrazine-2-carboxylic acid

Purity: 98+% CAS No.: 5424-01-1 Molecular Formula: C5H5N3O2 Formula Weight: 139.11 Melting Point: 204-206° dec. Beilstein Reference: 124835

Formel: C₅H₅N₃O₂
Molecular Weight: 139,11 g/mol
Schmelzpunkt: 204…206 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006141
CAS-Nummer: 5424-01-1
EINECS: 226-558-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR