3-Amino-2-pyrazincarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Amino-2-pyrazinecarboxylic acid
OS-7486-100G
OS-7486-25G
COBBOS-7486-100GEA
127
EUR
COBBOS-7486-100G
COBBOS-7486-25G
3-Amino-2-pyrazincarbonsäure
3-Aminopyrazine-2-carboxylic acid
Formel:
C₅H₅N₃O₂ Molecular Weight: 139,11 g/mol |
MDL:
MFCD00006141 CAS-Nummer: 5424-01-1 EINECS: 226-558-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...