2-Brom-5-methylpyrazin

Lieferant: COMBI-BLOCKS

PY-7723-1G PY-7723-5G
COBBPY-7723-1GEA 257 EUR
COBBPY-7723-1G COBBPY-7723-5G
2-Brom-5-methylpyrazin
2-Bromo-5-methylpyrazine
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD08705754
CAS-Nummer: 98006-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR