2-(Trifluormethyl)pyrazin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H33338.03 H33338.06
H33338.03EA 79.3 EUR
H33338.03 H33338.06
2-(Trifluormethyl)pyrazin ≥97%
2-(Trifluoromethyl)pyrazine

Purity: 97% CAS No.: 61655-67-2 Molecular Formula: C5H3F3N2 Formula Weight: 148.09 Boiling Point: 116-118° UN No.: 2810 MDL No.: MFCD06797734

Formel: C₅H₃F₃N₂
Molecular Weight: 148.09 g/mol
Siedepunkt: 116…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797734
CAS-Nummer: 61655-67-2
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR