2-Ethyl-3-(methylthio)pyrazin
Lieferant: FLUOROCHEM
002752-1G
002752-5G
FLUO002752-1GEA
31.1
EUR
FLUO002752-1G
FLUO002752-5G
2-Ethyl-3-(methylthio)pyrazin
2-Ethyl-3-(methylthio)pyrazine
Formel:
C₇H₁₀N₂S |
MDL:
MFCD00038027 CAS-Nummer: 72987-62-3 EINECS: 277-191-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...