Natrium-4-hydroxymercuribenzoat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
4-(Hydroxymercuri)benzoic acid sodium salt
55540-25G
55540-5G
SIAL55540-25GEA
426
EUR
SIAL55540-25G
SIAL55540-5G
Natrium-4-hydroxymercuribenzoat, Sigma-Aldrich®
Natrium-4-hydroxymercuribenzoat
Formel:
C₇H₅HgNaO₃ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
138-85-2 EINECS: 205-340-5 UN: 2025 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...