2-Acetyl-3-amino-5-phenylthiophen
Lieferant: FLUOROCHEM
CFF499.5
CFF499.25
017507-5G
017507-25G
CHMPCFF499.5EA
348
EUR
CHMPCFF499.5
CHMPCFF499.25
FLUO017507-5G
FLUO017507-25G
2-Acetyl-3-amino-5-phenylthiophen
2-Acetyl-3-amino-5-phenylthiophen
Formel:
C₁₂H₁₁NOS Molecular Weight: 217,29 g/mol |
MDL:
MFCD00068132 CAS-Nummer: 105707-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...