1-Chlor-4-fluor-2-iodbenzol

Lieferant: CHEMPUR

FL13273.POR 013273-25G 013273-5G
CHMPFL13273.POREA 0 EUR
CHMPFL13273.POR FLUO013273-25G FLUO013273-5G
1-Chlor-4-fluor-2-iodbenzol
1-Chloro-4-fluoro-2-iodobenzene
Formel: C₆H₃ClFI
Molecular Weight: 256,4439 g/mol
MDL: MFCD00042575
CAS-Nummer: 202982-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR