1-Chlor-4-fluor-3-iodbenzol
Lieferant: CHEMPUR
Synonyme:
4-Chloro-1-fluoro-2-iodobenzene
FL13274.POR
013274-25G
013274-5G
CHMPFL13274.POREA
0
EUR
CHMPFL13274.POR
FLUO013274-25G
FLUO013274-5G
1-Chlor-4-fluor-3-iodbenzol
1-Chloro-4-fluoro-3-iodobenzene
Formel:
C₆H₃ClFI Molecular Weight: 256,4400 g/mol |
MDL:
MFCD00672963 CAS-Nummer: 116272-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...