tert-Butylethylmalonat 95%
Lieferant: Thermo Scientific
Achtung
|
A11927.14
A11927.22
A11927.14EA
135
EUR
A11927.14
A11927.22
tert-Butylethylmalonat 95%
tert-Butylethylmalonat
Purity: 95%
CAS No.: 32864-38-3
Molecular Formula: C9H16O4
Formula Weight: 188.22
Flash Point: 87°(188°F)
Refractive Index: 1.4160
Boiling Point: 98-100°/22mm
Beilstein Reference: 1776681
MDL No.: MFCD00009193
Formel:
C₉H₁₆O₄ Molecular Weight: 188,22 g/mol Siedepunkt: 98…100 °C (22 mmHg) Dichte: 0,994 Flammpunkt: 87 °C (188 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009193 CAS-Nummer: 32864-38-3 EINECS: 251-269-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...