Decyltrimethylammoniumbromid, Sigma-Aldrich®
Lieferant: Merck
Synonyme: (1-Decyl)trimethylammonium bromide, N,N,N-Trimethyldecan-1-aminium bromide
30725-10G 30725-50G
SIAL30725-10GEA 63.5 EUR
SIAL30725-10G SIAL30725-50G
Decyltrimethylammoniumbromid, Sigma-Aldrich®
Decyltrimethylammonium bromide
Formel: C₁₃H₃₀BrN
Molecular Weight: 280.30 g/mol
MDL: MFCD00041973
CAS-Nummer: 2082-84-0
EINECS: 218-219-7