4-Brom-2-iodtoluol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2-iodo-1-methylbenzene

OT-0988-5G OT-0988-25G OT-0988-1G
COBBOT-0988-5GEA 185 EUR
COBBOT-0988-5G COBBOT-0988-25G COBBOT-0988-1G
4-Brom-2-iodtoluol
4-Brom-2-iodtoluol
Formel: C₇H₆BrI
Molecular Weight: 296,9300 g/mol
MDL: MFCD07780671
CAS-Nummer: 260558-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR