2,6-Dibrom-3-hydroxypyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dibromopyridin-3-ol

PY-7708-5G PY-7708-1G
COBBPY-7708-5GEA 310 EUR
COBBPY-7708-5G COBBPY-7708-1G
2,6-Dibrom-3-hydroxypyridin
2,6-Dibromo-3-hydroxypyridine
Formel: C₅H₃Br₂NO
Molecular Weight: 252,8900 g/mol
MDL: MFCD01318479
CAS-Nummer: 6602-33-1
EINECS: 229-548-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR