3,5-Dibrom-4-pyridon

Lieferant: COMBI-BLOCKS

HC-6784-5G HC-6784-1G
COBBHC-6784-5GEA 153 EUR
COBBHC-6784-5G COBBHC-6784-1G
3,5-Dibrom-4-pyridon
3,5-Dibrom-4-pyridon
Formel: C₅H₃Br₂NO
MDL: MFCD00082969
CAS-Nummer: 141375-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR