4-Brom-2-pyridon ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine

H32067.03 H32067.06
H32067.03EA 73.4 EUR
H32067.03 H32067.06
4-Brom-2-pyridon ≥97%
4-Brom-2-pyridon

Purity: 97% CAS No.: 36953-37-4 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 201-204° MDL No.: MFCD00234041

Formel: C₅H₄BrNO
Molecular Weight: 174,00 g/mol
Schmelzpunkt: 201…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234041
CAS-Nummer: 36953-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR