4-Brom-2-pyridon

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine

PY-7396-1G PY-7396-25G PY-7396-5G
COBBPY-7396-1GEA 25.9 EUR
COBBPY-7396-1G COBBPY-7396-25G COBBPY-7396-5G
4-Brom-2-pyridon
4-Brom-2-pyridon
Formel: C₅H₄BrNO
Molecular Weight: 174,00 g/mol
MDL: MFCD00234041
CAS-Nummer: 36953-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR