Methyltributylammoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tributylmethylammonium chloride, N,N-Dibutyl-N-methylbutan-1-aminium chloride
70444-100G-F
70444-500G-F
SIAL70444-100G-FEA
177
EUR
SIAL70444-100G-F
SIAL70444-500G-F
Methyltributylammoniumchlorid, Sigma-Aldrich®
Methyltributylammonium chloride
Formel:
C₁₃H₃₀ClN Molecular Weight: 235.84 g/mol |
MDL:
MFCD00011847 CAS-Nummer: 56375-79-2 EINECS: 260-135-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...