5-(Chloracetyl)oxindol

Lieferant: COMBI-BLOCKS

Synonyme: 5-(Chloroacetyl)-1,3-dihydro-2H-indol-2-one, 5-(2-Chloroacetyl)indolin-2-one

IN-0068-5G IN-0068-1G IN-0068-25G
COBBIN-0068-5GEA 114 EUR
COBBIN-0068-5G COBBIN-0068-1G COBBIN-0068-25G
5-(Chloracetyl)oxindol
5-(Chloracetyl)oxindol
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,6300 g/mol
MDL: MFCD04115734
CAS-Nummer: 65435-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR