5-(Chloracetyl)-6-chloroxindol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-5-(chloroacetyl)-1,3-dihydro-2H-indol-2-one

IN-0106-5G IN-0106-25G IN-0106-1G
COBBIN-0106-5GEA 196 EUR
COBBIN-0106-5G COBBIN-0106-25G COBBIN-0106-1G
5-(Chloracetyl)-6-chloroxindol
5-(Chloracetyl)-6-chloroxindol
Formel: C₁₀H₇Cl₂NO₂
MDL: MFCD04115735
CAS-Nummer: 118307-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR