Cyclopropylessigsäure
Lieferant: FLUOROCHEM
003710-1G
003710-5G
003710-25G
FLUO003710-1GEA
60.9
EUR
FLUO003710-1G
FLUO003710-5G
FLUO003710-25G
Cyclopropylessigsäure
Cyclopropylessigsäure
Formel:
C₅H₈O₂ Molecular Weight: 100.12 g/mol |
MDL:
MFCD00041544 CAS-Nummer: 5239-82-7 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...