2-(1,1,2,2-Tetrafluorethoxy)anilin
Lieferant: FLUOROCHEM
010529-25G
010529-5G
FLUO010529-25GEA
206
EUR
FLUO010529-25G
FLUO010529-5G
2-(1,1,2,2-Tetrafluorethoxy)anilin
2-(1,1,2,2-Tetrafluorethoxy)anilin
Formel:
C₈H₇F₄NO |
MDL:
MFCD03094306 CAS-Nummer: 35295-34-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...