3-(1,1,2,2-Tetrafluorethoxy)anilin

Lieferant: FLUOROCHEM

010530-25G 010530-100G 010530-5G
FLUO010530-25GEA 0 EUR
FLUO010530-25G FLUO010530-100G FLUO010530-5G
3-(1,1,2,2-Tetrafluorethoxy)anilin
3-(1,1,2,2-Tetrafluorethoxy)anilin
Formel: C₈H₇F₄NO
MDL: MFCD00042457
CAS-Nummer: 831-75-4
EINECS: 212-610-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR