O-Methylhydroxylamin Hydrochlorid 25 - 30% in wässriger Lösung, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

225517-100G 225517-25G
SIAL225517-100GEA 54.1 EUR
SIAL225517-100G SIAL225517-25G
O-Methylhydroxylamin Hydrochlorid 25 - 30% in wässriger Lösung, Sigma-Aldrich®
O-Methylhydroxylamin Hydrochlorid
Formel: CH₆ClNO
Molecular Weight: 83,515 g/mol
MDL: MFCD00012951
CAS-Nummer: 593-56-6
EINECS: 209-798-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR