N-Acetyl-L-methionin

Lieferant: BACHEM BIOCHEMICA

Synonyme: N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH

4003668.0005 4003668.0025
BACE4003668.0005EA 0 EUR
BACE4003668.0005 BACE4003668.0025
N-Acetyl-L-methionin
N-Acetyl-L-methionin
Formel: C₇H₁₃NO₃S
Molecular Weight: 191.2 g/mol
MDL: MFCD00064441
CAS-Nummer: 65-82-7
EINECS: 200-617-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR