N-Acetyl-L-methionin, Sigma-Aldrich®
Lieferant: Merck
Synonyme: N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH
MQ100
01310-25G 01310-5G
SIAL01310-25GEA 366 EUR
SIAL01310-25G SIAL01310-5G
N-Acetyl-L-methionin, Sigma-Aldrich®
N-Acetyl-L-methionin
Formel: C₇H₁₃NO₃S
Molecular Weight: 191.2 g/mol
MDL: MFCD00064441
CAS-Nummer: 65-82-7
EINECS: 200-617-7