Dimethylglyoxim Dinatriumsalz Octahydrat analytisches Reagens
Lieferant: Bernd Kraft
20473.0600
20473.2600
20473.3600
KRAF20473.0600EA
143
EUR
KRAF20473.0600
KRAF20473.2600
KRAF20473.3600
Dimethylglyoxim Dinatriumsalz Octahydrat analytisches Reagens
Dimethylglyoxim Dinatriumsalz Octahydrat
Formel:
C₄H₆N₂Na₂O₂·8H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150288 CAS-Nummer: 75006-64-3 EINECS: 262-523-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...