2,4-Dichlorphenyloxamidsäureethylester

Lieferant: BIONET RESEARCH

11E-069.500 11E-069.1 11E-069.5 11E-069.10
BIOT11E-069.500EA 0 EUR
BIOT11E-069.500 BIOT11E-069.1 BIOT11E-069.5 BIOT11E-069.10
2,4-Dichlorphenyloxamidsäureethylester
2,4-Dichlorphenyloxamidsäureethylester
Formel: C₁₀H₉Cl₂NO₃
CAS-Nummer: 15313-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR