Ethylmalonylchlorid 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Ethyl Malonyl Chloride

A15616.06 A15616.14 A15616.22
A15616.06EA 55.9 EUR
A15616.06 A15616.14 A15616.22
Ethylmalonylchlorid 95%
Ethylmalonylchlorid

Purity: 95% CAS No.: 36239-09-5 Molecular Formula: C5H7ClO3 Formula Weight: 150.56 Storage Temperature: 2°C to 8°C Flash Point: 67°(152°F) Refractive Index: 1.4290 Boiling Point: 83°/22mm Beilstein Reference: 636215 UN No.: 3265 MDL No.: MFCD00000736

Formel: C₅H₇ClO₃
Molecular Weight: 150,56 g/mol
Siedepunkt: 83 °C (22 mmHg)
Dichte: 1,189
Flammpunkt: 67 °C (152 °F)
MDL: MFCD00000736
CAS-Nummer: 36239-09-5
EINECS: 252-934-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR