Ethylmalonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl Malonyl Chloride

M-Clarity™ Qualitätsstufe = MQ200

163872-25G 163872-5G
SIAL163872-25GEA 315 EUR
SIAL163872-25G SIAL163872-5G
Ethylmalonylchlorid, Sigma-Aldrich®
Ethylmalonylchlorid
Formel: C₅H₇ClO₃
Molecular Weight: 150,56 g/mol
MDL: MFCD00000736
CAS-Nummer: 36239-09-5
EINECS: 252-934-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR