Methylmalonylchlorid

Lieferant: CHEMPUR

Synonyme: Methyl 3-chloro-3-oxopropanoate

FL13167.POR 013167-25G
CHMPFL13167.POREA 0 EUR
CHMPFL13167.POR FLUO013167-25G
Methylmalonylchlorid
Methylmalonylchlorid
Formel: C₄H₅ClO₃
Molecular Weight: 136,53 g/mol
MDL: MFCD00013657
CAS-Nummer: 37517-81-0
EINECS: 253-540-6
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR