Methylmalonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3-chloro-3-oxopropanoate

M-Clarity™ Qualitätsstufe = MQ200

164011-25G 164011-5G
SIAL164011-25GEA 160 EUR
SIAL164011-25G SIAL164011-5G
Methylmalonylchlorid, Sigma-Aldrich®
Methylmalonylchlorid
Formel: C₄H₅ClO₃
Molecular Weight: 136,53 g/mol
MDL: MFCD00013657
CAS-Nummer: 37517-81-0
EINECS: 253-540-6
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR