(2-Methoxyethoxy)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(2-Methoxyethoxy)acetic acid

M-Clarity™ Qualitätsstufe = MQ100

407011-1L 407011-250ML
SIAL407011-1LEA 320 EUR
SIAL407011-1L SIAL407011-250ML
(2-Methoxyethoxy)essigsäure, Sigma-Aldrich®
(2-Methoxyethoxy)essigsäure
Formel: C₅H₁₀O₄
Molecular Weight: 134,13 g/mol
MDL: MFCD00044097
CAS-Nummer: 16024-56-9
EINECS: 240-161-6
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR