4-Fluorphenyloxamidsäureethylester

Lieferant: BIONET RESEARCH

Synonyme: Ethyl 2-((4-fluorophenyl)amino)-2-oxoacetate, Ethyl 2-(4-fluoroanilino)-2-oxoacetate

2F-079.500 2F-079.1 2F-079.5 2F-079.10
BIOT2F-079.500EA 0 EUR
BIOT2F-079.500 BIOT2F-079.1 BIOT2F-079.5 BIOT2F-079.10
4-Fluorphenyloxamidsäureethylester
4-Fluorphenyloxamidsäureethylester
Formel: C₁₀H₁₀FNO₃
Molecular Weight: 211,1897 g/mol
MDL: MFCD00119045
CAS-Nummer: 69065-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR