n-Butylisocyanid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

133280-1G 133280-5G
SIAL133280-1GEA 99.8 EUR
SIAL133280-1G SIAL133280-5G
n-Butylisocyanid, Sigma-Aldrich®
n-Butylisocyanid
Formel: C₅H₉N
Molecular Weight: 83,132 g/mol
MDL: MFCD00000008
CAS-Nummer: 2769-64-4
EINECS: 220-458-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR