Diethyl(2-cyanethyl)malonat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Diethyl 2-(2-cyanoethyl)malonate
A11567.09
A11567.14
A11567.18
A11567.09EA
25.7
EUR
A11567.09
A11567.14
A11567.18
Diethyl(2-cyanethyl)malonat ≥98%
Diethyl(2-cyanethyl)malonat
Purity: 98+%
CAS No.: 17216-62-5
Molecular Formula: C10H15NO4
Formula Weight: 213.23
Flash Point:
Formel:
C₁₀H₁₅NO₄ Molecular Weight: 213.23 g/mol Siedepunkt: 115…117 °C (2 mmHg) Dichte: 1.079 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001966 CAS-Nummer: 17216-62-5 EINECS: 241-260-7 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...