(4-Nitrophenyl)glyoxylsäure

Lieferant: CHEMPUR

Synonyme: 2-(4-Nitrophenyl)-2-oxoacetic acid

JFD01270.250 JFD01270.1
CHMPJFD01270.250EA 0 EUR
CHMPJFD01270.250 MAYBJFD01270.1
(4-Nitrophenyl)glyoxylsäure
(4-Nitrophenyl)glyoxylsäure
Formel: C₈H₅NO₅
Molecular Weight: 195,1300 g/mol
MDL: MFCD00051744
CAS-Nummer: 14922-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR