(1-Amino-2-chlorethyliden)malononitril

Lieferant: BIONET RESEARCH

5C-001.500 5C-001.1 5C-001.5 5C-001.10
BIOT5C-001.500EA 0 EUR
BIOT5C-001.500 BIOT5C-001.1 BIOT5C-001.5 BIOT5C-001.10
(1-Amino-2-chlorethyliden)malononitril
(1-Amino-2-chlorethyliden)malononitril
Formel: C₅H₄ClN₃
CAS-Nummer: 118645-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR