(1-Amino-2-phenoxyethyliden)malononitril

Lieferant: BIONET RESEARCH (CHEMPUR)

BIOT7C-013.1EA 0 EUR
BIOT7C-013.1 BIOT7C-013.10 BIOT7C-013.500 BIOT7C-013.5
(1-Amino-2-phenoxyethyliden)malononitril
(1-Amino-2-phenoxyethyliden)malononitril
Formel: C₁₁H₉N₃O
CAS-Nummer: 118645-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR