Ethyl-3-isocyanatobenzoat ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-(Ethoxycarbonyl)phenyl isocyanate
L09495.03
L09495.06
L09495.14
L09495.03EA
35.8
EUR
L09495.03
L09495.06
L09495.14
Ethyl-3-isocyanatobenzoat ≥97%
Ethyl-3-isocyanatobenzoat
Purity: 97%
CAS No.: 67531-68-4
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₁₀H₉NO₃ Molecular Weight: 191,19 g/mol Siedepunkt: 213 °C Dichte: 1,154 Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00013865 CAS-Nummer: 67531-68-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...