Methyl-3-isocyanatobenzoat ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Methyl3-isocyanatobenzoate, 3-(Methoxycarbonyl)phenyl isocyanate
L18709.06
L18709.03
L18709.06EA
221
EUR
L18709.06
L18709.03
Methyl-3-isocyanatobenzoat ≥97%
Methyl-3-isocyanatobenzoat
Purity: 97%
CAS No.: 41221-47-0
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Flash Point:
Formel:
C₉H₇NO₃ Molecular Weight: 177,16 g/mol Schmelzpunkt: 35…38 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037077 CAS-Nummer: 41221-47-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...